Skip to Content
MilliporeSigma
All Photos(1)

Documents

38545

Sigma-Aldrich

Bis[2-(4-azidosalicylamido)ethyl] disulfide

≥95.0% (HPLC)

Synonym(s):

BASED, N, N′-(Dithiobis-ethylene)bis(4-azido-2-hydroxy-benzamide), N,N′-Bis(4-azidosalicoyl)cystamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18N8O4S2
CAS Number:
Molecular Weight:
474.52
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥95.0% (HPLC)

storage temp.

2-8°C

SMILES string

Oc1cc(ccc1C(=O)NCCSSCCNC(=O)c2ccc(cc2O)N=[N+]=[N-])N=[N+]=[N-]

InChI

1S/C18H18N8O4S2/c19-25-23-11-1-3-13(15(27)9-11)17(29)21-5-7-31-32-8-6-22-18(30)14-4-2-12(24-26-20)10-16(14)28/h1-4,9-10,27-28H,5-8H2,(H,21,29)(H,22,30)

InChI key

QQZOUYFHWKTGEY-UHFFFAOYSA-N

Application

Reagent for cross linking of proteins; the cross link can be cleaved

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steffen Schumann et al.
IEEE transactions on bio-medical engineering, 62(11), 2665-2673 (2015-06-10)
In the following, we will present a newly developed X-ray calibration phantom and its integration for 2-D/3-D pelvis reconstruction and subsequent automatic cup planning. Two different planning strategies were applied and evaluated with clinical data. Two different cup planning methods
Siheng Chen et al.
IEEE transactions on image processing : a publication of the IEEE Signal Processing Society (2019-12-14)
We propose a deep autoencoder with graph topology inference and filtering to achieve compact representations of unorganized 3D point clouds in an unsupervised manner. Many previous works discretize 3D points to voxels and then use lattice-based methods to process and
Bingqing Xie et al.
Journal of computational biology : a journal of computational molecular cell biology, 22(4), 313-323 (2015-04-07)
Identifying high-confidence candidate genes that are causative for disease phenotypes, from the large lists of variations produced by high-throughput genomics, can be both time-consuming and costly. The development of novel computational approaches, utilizing existing biological knowledge for the prioritization of
J Rappsilber et al.
Analytical chemistry, 72(2), 267-275 (2000-02-05)
Most cellular functions are performed by multi-protein complexes. The identity of the members of such complexes can now be determined by mass spectrometry. Here we show that mass spectrometry can also be used in order to define the spatial organization
Rui Zhang et al.
Clinical chemistry, 65(6), 761-770 (2019-03-16)
Noninvasive prenatal testing (NIPT) based on cell-free DNA (cfDNA) is widely used. However, biomimetic quality control materials that have properties identical to clinical samples and that are applicable to a wide range of methodologies are still not available to support

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service