Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14955

Sigma-Aldrich

Iris 7G-WS carboxylic acid

Synonym(s):

2-{2-{2-[4-(4-Carboxybutyrylamino)phenylthio]-3-{2-[1-(hex-5-ynyl)-1,3-dihydro-3,3-dimethyl-5-sulfoindol-2-ylidene]ethylidene}cyclohex-1-enyl}vinyl}-1-(hex-5-ynyl)-3,3-dimethyl-3H-indolium-5-sulfonate monopotassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H58KN3O9S3
CAS Number:
Molecular Weight:
1016.34
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

fluorescence

λex 804 nm; λem 823 nm in 0.1 M phosphate pH 7.0

storage temp.

−20°C

SMILES string

[K+].CC1(C)/C(=C\C=C2/CCCC(\C=C\C3=[N+](CCCCC#C)c4ccc(cc4C3(C)C)S([O-])(=O)=O)=C2Sc5ccc(NC(=O)CCCC(O)=O)cc5)N(CCCCC#C)c6ccc(cc16)S([O-])(=O)=O

InChI

1S/C53H59N3O9S3.K/c1-7-9-11-13-33-55-45-29-27-41(67(60,61)62)35-43(45)52(3,4)47(55)31-21-37-17-15-18-38(51(37)66-40-25-23-39(24-26-40)54-49(57)19-16-20-50(58)59)22-32-48-53(5,6)44-36-42(68(63,64)65)28-30-46(44)56(48)34-14-12-10-8-2;/h1-2,21-32,35-36H,9-20,33-34H2,3-6H3,(H3-,54,57,58,59,60,61,62,63,64,65);/q;+1/p-1

InChI key

GLVAQGAOXASSSF-UHFFFAOYSA-M

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Principle

The dye can be activated at the carboxy group for coupling purposes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Radhika A Varier et al.
The EMBO journal, 29(23), 3967-3978 (2010-10-19)
Histone methylation patterns are correlated with eukaryotic gene transcription. High-affinity binding of the plant homeodomain (PHD) of TFIID subunit TAF3 to trimethylated lysine-4 of histone H3 (H3K4me3) is involved in promoter recruitment of this basal transcription factor. Here, we show
Chiara A Bertolino et al.
Journal of fluorescence, 16(2), 221-225 (2006-05-03)
A series of novel functionalized, water-soluble, pH-unsensitive, highly photostable heptamethine cyanine dyes (HCDs) has been synthesized. The aim of the synthesis was to obtain novel effective probes for fluorescence detection in the near infrared. Synthesis and characterization of a special

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service