Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

61167

Sigma-Aldrich

Copper(II) carbonate basic

purum p.a., ≥95% (RT)

Synonym(s):

Cupric carbonate basic

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuCO3 · Cu(OH)2
CAS Number:
Molecular Weight:
221.12
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥95% (RT)
form:
powder

grade

purum p.a.

Assay

≥95% (RT)

form

powder

anion traces

chloride (Cl-): ≤1000 mg/kg
sulfate (SO42-): ≤200 mg/kg

cation traces

Ca: ≤5000 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Fe: ≤100 mg/kg
K: ≤100 mg/kg
Na: ≤5000 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤1000 mg/kg

SMILES string

[Cu++].O[Cu]O.[O-]C([O-])=O

InChI

1S/CH2O3.2Cu.2H2O/c2-1(3)4;;;;/h(H2,2,3,4);;;2*1H2/q;2*+2;;/p-4

InChI key

ZMMDPCMYTCRWFF-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Copper(II) carbonate basic (Cupric carbonate basic) is a basic copper(II) salt.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Preparation and characterization of uniform particles of pure and coated metallic copper.
Hsu WP, et al.
Powder Technology, 63(3), 265-275 (1990)
Cathodic reduction and infrared reflectance spectroscopy of basic copper (II) salts on copper substrate.
Malvault JY, et al.
J. Appl. Electrochem., 25(9), 841-845 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service