Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

31253

Sigma-Aldrich

Potassium hexacyanoferrate(III)

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99%

Synonym(s):

Potassium ferricyanide, Red prussiate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K3Fe(CN)6
CAS Number:
Molecular Weight:
329.24
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24

grade

ACS reagent
puriss. p.a.

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

Assay

≥99%

form

crystalline

technique(s)

titration: suitable

impurities

≤0.02% hexacyanoferrate(II) ([Fe(CN)6]4-)

pH

6-9 (25 °C, 329 g/L)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤5 mg/kg
Cd: ≤5 mg/kg
Co: ≤50 mg/kg
Cu: ≤10 mg/kg
Na: ≤200 mg/kg
Ni: ≤10 mg/kg
Pb: ≤20 mg/kg
Zn: ≤5 mg/kg

SMILES string

[K+].[K+].[K+].N#C[Fe-3](C#N)(C#N)(C#N)(C#N)C#N

InChI

1S/6CN.Fe.3K/c6*1-2;;;;/q;;;;;;-3;3*+1

InChI key

MIMJFNVDBPUTPB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Potassium hexacyanoferrate(III) (Potassium ferricyanide) is an alkali hexacyanoferrate. On heating, it undergoes decomposition to afford yellow-brown hexacyanoiron(III) acid. It can be prepared by the anodic oxidation of K4[Fe(CN)6].

Application

Potassium hexacyanoferrate(III) may be employed as an oxidizing agent during the fabrication of glucose conductometric biosensor.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2

Supplementary Hazards

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eagleson M.
Concise Encyclopedia Chemistry, 291-291 (1994)
Alemnew Geto et al.
Environment international, 129, 400-407 (2019-06-04)
Bentazone is one of the most problematic pesticides polluting groundwater resources. It is on the list of pesticides that are mandatory to analyze at water work controls. The current pesticide measuring approach includes manual water sampling and time-consuming chromatographical quantification
Glucose conductometric biosensor with potassium hexacyanoferrate (III) as an oxidizing agent.
Dzyadevich SV, et al.
Analytica Chimica Acta, 374(1), 11-18 (1998)
Islem Gandouzi et al.
Materials (Basel, Switzerland), 12(7) (2019-04-14)
Pyoverdine is a fluorescent siderophore produced by Pseudomonas aeruginosa that can be considered as a detectable marker in nosocomial infections. The presence of pyoverdine in water can be directly linked to the presence of the P. aeruginosa, thus being a
Mark A Jordan et al.
Water research, 47(2), 841-849 (2012-12-04)
Representative and fast monitoring of wastewater influent and effluent biochemical oxygen demand (BOD) is an elusive goal for the wastewater industry and regulatory bodies alike. The present study describes a suitable assay, which incorporates activated sludge as the biocatalyst and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service