Skip to Content
MilliporeSigma
All Photos(1)

Documents

30103

Sigma-Aldrich

Hydrofluoric acid

puriss. p.a., reag. ISO, reag. Ph. Eur., ≥40%

Synonym(s):

HF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HF
CAS Number:
Molecular Weight:
20.01
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

vapor density

1.27 (vs air)

vapor pressure

25 mmHg ( 20 °C)

grade

puriss. p.a.

Assay

≥40%

form

liquid

impurities

≤0.005% hexafluorosilic acid (H2SiF6)

ign. residue

≤0.0005% (as SO4)

density

1.15 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤1 mg/kg
phosphate (PO43-): ≤0.5 mg/kg
sulfate (SO42-): ≤2 mg/kg
sulfite (SO32-): ≤2 mg/kg

cation traces

Ag: ≤0.02 mg/kg
Al: ≤0.05 mg/kg
As: ≤0.05 mg/kg
Ba: ≤0.1 mg/kg
Be: ≤0.02 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.01 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.2 mg/kg
Ge: ≤0.05 mg/kg
K: ≤0.1 mg/kg
Li: ≤0.02 mg/kg
Mg: ≤0.2 mg/kg
Mn: ≤0.05 mg/kg
Mo: ≤0.05 mg/kg
Na: ≤0.2 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.05 mg/kg
Sr: ≤0.02 mg/kg
Ti: ≤0.1 mg/kg
Tl: ≤0.05 mg/kg
V: ≤0.05 mg/kg
Zn: ≤0.05 mg/kg
Zr: ≤0.1 mg/kg

SMILES string

[H+].[F-]

InChI

1S/FH/h1H

InChI key

KRHYYFGTRYWZRS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly-ordered TiO2 nanotube arrays up to 220μm in length: use in water photoelectrolysis and dye-sensitized solar cells.
Shankar K, et al.
Nanotechnology, 18(6), 065707-065707 (2007)
Francesca Monticelli et al.
Dental materials : official publication of the Academy of Dental Materials, 22(7), 602-609 (2005-11-18)
To verify the influence of different etching procedures of the post-surface on microtensile bond strength values between fiber posts and composite core materials. 60 DT Light Posts were divided into 10 subgroups using five different chemical surface treatments and two
Anatase TiO2 nanosheet: an ideal host structure for fast and efficient lithium insertion/extraction.
Chen JS and Lou XW.
Electrochemical Communications, 11(12), 2332-2335 (2009)
Eagleson M.
Concise Encyclopedia Chemistry, 508-508 (1994)
Nicolas Vodovar et al.
JACC. Heart failure, 3(8), 629-636 (2015-08-08)
This study sought to hypothesize that elevated B-type natriuretic peptide (BNP) could act as an endogenous neprilysin inhibitor. A hallmark of acute decompensated heart failure (ADHF) is the overproduction of natriuretic peptides (NPs) by stretched cardiomyocytes. Various strategies have been

Articles

Professor Gogotsi and Dr. Shuck introduce MXenes: a promising family of two-dimensional materials with a unique combination of high conductivity, hydrophilicity, and extensive tunability.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service