Skip to Content
MilliporeSigma
All Photos(1)

Documents

13206

Sigma-Aldrich

Potassium permanganate

puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
KMnO4
CAS Number:
Molecular Weight:
158.03
EC Number:
MDL number:
UNSPSC Code:
12352312
PubChem Substance ID:

grade

puriss.

Assay

99-100.5%

form

solid

quality

meets analytical specification of Ph. Eur., BP, USP

reaction suitability

reagent type: oxidant

impurities

residual solvents, complies

loss

≤0.2% loss on drying, 105 °C

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤300 mg/kg

suitability

complies for appearance of solution

SMILES string

[K+].[O-][Mn](=O)(=O)=O

InChI

1S/K.Mn.4O/q+1;;;;;-1

InChI key

VZJVWSHVAAUDKD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Suitable for heterogeneous oxidations.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 2 - Repr. 2 - Skin Corr. 1 - STOT RE 2 Inhalation

Target Organs

Brain

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Millicent G Managa et al.
Molecules (Basel, Switzerland), 25(6) (2020-03-19)
Chinese cabbage (Brassica rapa L. subsp. chinensis) and Nightshade (Solanum retroflexum are popular traditional leafy vegetables consumed predominantly by rural Africans. Sun drying is adopted as a traditional method of postharvest preservation to store theses leaves during off seasons. The
George Trakakis et al.
Materials (Basel, Switzerland), 13(19) (2020-10-01)
High volume fraction carbon nanotube (CNT) composites (7.5-16% vol.) were fabricated by the impregnation of CNT buckypapers into epoxy resin. To enhance the interfacial reaction with the epoxy resin, the CNTs were modified by two different treatments, namely, an epoxidation
Arnaud Beduneau et al.
PloS one, 4(2), e4343-e4343 (2009-02-03)
We posit that the same mononuclear phagocytes (MP) that serve as target cells and vehicles for a host of microbial infections can be used to improve diagnostics and drug delivery. We also theorize that physical and biological processes such as
Christopher H Gibbons et al.
Neurology, 72(17), 1479-1486 (2009-04-29)
To evaluate a novel method to quantify the density of nerve fibers innervating sweat glands in healthy control and diabetic subjects, to compare the results to an unbiased stereologic technique, and to identify the relationship to standardized physical examination and
Jessica M Terry et al.
Talanta, 99, 1051-1056 (2012-09-13)
We have explored the chemiluminescence response of amino acids and related compounds (including the tripeptide glutathione, and disulfides: cystine, homocystine and glutathione disulfide) with several new adaptations of the permanganate and tris(2,2'-bipyridine)ruthenium(III) ([Ru(bipy)(3)](3+)) reagents and a recently developed colloidal manganese(IV)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service