PHR3618
Valacyclovir Related Compound F
certified reference material, pharmaceutical secondary standard
Synonym(s):
L-Valine 2-hydroxyethyl ester p-toluenesulfonate salt
About This Item
Recommended Products
grade
certified reference material
pharmaceutical secondary standard
Quality Level
Agency
traceable to USP 1707883
CofA
current certificate can be downloaded
packaging
pkg of 50 mg
application(s)
pharmaceutical
storage temp.
2-8°C
SMILES string
CC(C)[C@H](N)C(=O)OCCO.Cc1ccc(cc1)S(O)(=O)=O
InChI
1S/C7H15NO3.C7H8O3S/c1-5(2)6(8)7(10)11-4-3-9;1-6-2-4-7(5-3-6)11(8,9)10/h5-6,9H,3-4,8H2,1-2H3;2-5H,1H3,(H,8,9,10)/t6-;/m0./s1
InChI key
KIQNQGPNCRYWKY-RGMNGODLSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Analysis Note
Other Notes
Footnote
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service