Skip to Content
MilliporeSigma
All Photos(1)

Documents

IRMM540R

Oxide glass

IRMM®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

IRMM®

application(s)

pharmaceutical (small molecule)

format

matrix material

Looking for similar products? Visit Product Comparison Guide

Physical form

discs, 15 mm diameter

Analysis Note

For more information please see:
IRMM540R

Legal Information

IRMM is a registered trademark of European Commission

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service