Skip to Content
MilliporeSigma
All Photos(1)

Documents

92775

Sigma-Aldrich

Trimethylsilyl 3-(trimethylsilyl)propynoate

purum, ≥97.0% (GC)

Synonym(s):

3-(Trimethylsilyl)propynoic acid trimethylsilyl ester, Trimethylsilyl (trimethylsilyl)acetylenecarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiC≡CCO2Si(CH3)3
CAS Number:
Molecular Weight:
214.41
MDL number:
UNSPSC Code:
12352100

grade

purum

Assay

≥97.0% (GC)

refractive index

n20/D 1.44 (lit.)

bp

64-65 °C/1.5 mmHg (lit.)

mp

~25 °C

shipped in

wet ice

storage temp.

−20°C

SMILES string

C[Si](C)(C)OC(=O)C#C[Si](C)(C)C

Looking for similar products? Visit Product Comparison Guide

Other Notes

Esters of trimethylsilyl-propinoic acid are useful reagents for [2+2+2]-cycloadditions and similar reactions

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K.P.C. Vollhardt
Angewandte Chemie (International Edition in English), 96, 525-525 (1984)
K.C. Nicolaou et al.
Journal of the Chemical Society. Chemical Communications, 421-421 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service