Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

87254

Supelco

Tetraethylphosphonium tetrafluoroborate

Selectophore, ≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2)4P(BF4)
CAS Number:
Molecular Weight:
234.02
Beilstein:
3730026
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
Pricing and availability is not currently available.

product line

Selectophore

Assay

≥98.0% (T)
≥98.0%

mp

300-302 °C

SMILES string

F[B-](F)(F)F.CC[P+](CC)(CC)CC

Other Notes

Novel organic electrolyte with high conductivity in high energy density electric double-layer capacitors (EDLC)[1]

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T. Morimoto et al.
Proc. Electrochem. Soc., 93/23, 49-49 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service