Skip to Content
MilliporeSigma
All Photos(1)

Documents

77232

Sigma-Aldrich

Perchloric acid

puriss. p.a., ACS reagent, ≥60% (T)

Synonym(s):

PCA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HClO4
CAS Number:
Molecular Weight:
100.46
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

ACS reagent
puriss. p.a.

concentration

≥60% (T)

impurities

≤0.00005% free chlorine (Cl)
≤0.001% total nitrogen (N)

ign. residue

≤0.003% (as SO4)

anion traces

chlorate (ClO3-): ≤10 mg/kg
chloride (Cl-): ≤3 mg/kg
phosphate, silicate (as SiO2): ≤5 mg/kg
sulfate (SO42-): ≤10 mg/kg

cation traces

Ag: ≤0.1 mg/kg
Al: ≤0.05 mg/kg
As: ≤0.05 mg/kg
Ba: ≤0.02 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.05 mg/kg
Cu: ≤0.1 mg/kg
Fe: ≤1 mg/kg
K: ≤0.1 mg/kg
Li: ≤0.02 mg/kg
Mg: ≤0.5 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.05 mg/kg
Ni: ≤0.1 mg/kg
Pb: ≤0.05 mg/kg
Sr: ≤0.02 mg/kg
Tl: ≤0.05 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

OCl(=O)(=O)=O

InChI

1S/ClHO4/c2-1(3,4)5/h(H,2,3,4,5)

InChI key

VLTRZXGMWDSKGL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 1 - Skin Corr. 1A - STOT RE 2

Target Organs

Thyroid

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

He-Ping Zhao et al.
Environmental science & technology, 47(3), 1565-1572 (2013-01-10)
We evaluated a strategy for achieving complete reduction of perchlorate (ClO(4)(-)) in the presence of much higher concentrations of sulfate (SO(4)(2-)) and nitrate (NO(3)(-)) in a hydrogen-based membrane biofilm reactor (MBfR). Full ClO(4)(-) reduction was achieved by using a two-stage
J R Anoop Raj et al.
Environmental technology, 34(5-8), 841-852 (2013-07-11)
The present work is a laboratory-scale study of perchlorate degradation using Proteobacterium ARJR SMBS in a stirred tank bioreactor (STBR). Anaerobically grown cultures of ARJR SMBS exposed to a variety of ClO4(-) levels within the range 30 to 150 mg
Craig Steinmaus et al.
Environmental research, 123, 17-24 (2013-03-12)
Perchlorate, thiocyanate, and low iodine intake can all decrease iodide intake into the thyroid gland. This can reduce thyroid hormone production since iodide is a key component of thyroid hormone. Previous research has suggested that each of these factors alone
Lunar and planetary science conference. Pesky perchlorates all over Mars.
Richard A Kerr
Science (New York, N.Y.), 340(6129), 138-138 (2013-04-13)
Yiqiong Yang et al.
Water environment research : a research publication of the Water Environment Federation, 85(4), 331-339 (2013-05-24)
Perchlorate is widely known as an inorganic endocrine disruptor. In this study, MgAl-CO3 layered double hydroxides with different Mg/Al molar ratios were prepared using a coprecipitation method and followed by a calcination process at a temperature range of 300 to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service