Skip to Content
MilliporeSigma
All Photos(1)

Documents

74736

Supelco

N-Octadecanoyl-Nile blue

for fluorescence, ≥98.0% (TLC)

Synonym(s):

Chromoionophore I, 3-Octadecanoylimino-7-(diethylamino)-1,2-benzophenoxazine, 9-(Diethylamino)-5-(octadecanoylimino)-5H-benzo[a]phenoxazine, N-Octadecanoyl-Nile blue, ETH 5294

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H53N3O2
CAS Number:
Molecular Weight:
583.85
Beilstein:
3576427
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

grade

for fluorescence

Assay

≥98.0% (TLC)

mp

91-93 °C (lit.)

solubility

DMF: soluble
DMSO: soluble
alcohols: soluble

fluorescence

λex 614 nm; λem 663 nm (protonated)

SMILES string

CCCCCCCCCCCCCCCCCC(=O)\N=C1/C=C2Oc3cc(ccc3N=C2c4ccccc14)N(CC)CC

InChI

1S/C38H53N3O2/c1-4-7-8-9-10-11-12-13-14-15-16-17-18-19-20-25-37(42)39-34-29-36-38(32-24-22-21-23-31(32)34)40-33-27-26-30(28-35(33)43-36)41(5-2)6-3/h21-24,26-29H,4-20,25H2,1-3H3/b39-34+

InChI key

MYWBZQJIXVRCJC-JQJRJOTLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Application

Assay of K+ and of Ca2+ activity with solvent polymeric optode membrane based on ETH 5294 and potassium ionophore I (valinomycin)

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

comparable product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service