Skip to Content
MilliporeSigma
All Photos(1)

Documents

68437

Supelco

Kaempferol 3-glucoside

analytical standard

Synonym(s):

Kaempferol 3-β-D-glucopyranoside, 3,4′,5,7-Tetrahydroxyflavone 3-glucoside, 3-(β-D-Glucopyranosyloxy)-5,7-dihydroxy-2-(4-methoxyphenyl)-4H-1-benzopyran-4-one, 3-Glucosylkaempferol, Astragalin, Kaempferol 3-glucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20O11
CAS Number:
Molecular Weight:
448.38
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥90.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤12.0% water

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@@H](OC2=C(Oc3cc(O)cc(O)c3C2=O)c4ccc(O)cc4)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C21H20O11/c22-7-13-15(26)17(28)18(29)21(31-13)32-20-16(27)14-11(25)5-10(24)6-12(14)30-19(20)8-1-3-9(23)4-2-8/h1-6,13,15,17-18,21-26,28-29H,7H2/t13-,15-,17+,18-,21+/m1/s1

InChI key

JPUKWEQWGBDDQB-QSOFNFLRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Kaempferol 3-glucoside is a flavonoid glycoside that occurs naturally in Cleome and Capparis species of plants.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Similar kaempferol glycosides reduce food intake and corresponding weight gain in Wistar rats.The glucoside was not tested.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical characterization of three accessions of Brassica juncea L. extracts from different plant tissues
Oulad El Majdoub Y, et al.
Molecules (Basel) (2020)
Flavonoids of four Cleome and three Capparis species
Sharaf M, et al.
Biochemical Systematics and Ecology, 25, 161-166 (1997)
Determination of the metabolite content of Brassica juncea cultivars using comprehensive two-dimensional liquid chromatography coupled with a photodiode array and mass spectrometry detection
Arena K, et al.
Molecules (Basel), 25(5), 1235-1235 (2020)
Min Ke et al.
Bio-medical materials and engineering, 22(1-3), 113-119 (2012-07-07)
Diabetic retinopathy (DR) is a severe complication of diabetes mellitus (DM) and often causes vision loss or even blindness. Vascular endothelial growth factor (VEGF) in the retina, which is mainly derived from Müller cells, is a crucial biological factor in
Xianghui He et al.
Planta medica, 77(17), 1950-1957 (2011-06-16)
Cuscuta chinensis and Cuscuta australis, the two botanical sources of the Chinese herbal medicine Tu-Si-Zi, were distinguished from each other based on qualitative and quantitative chemical analysis. By HPLC‑DAD‑MS, a total of 36 compounds were characterized from these two Cuscuta

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service