Skip to Content
MilliporeSigma
All Photos(1)

Documents

63761

Sigma-Aldrich

3-Mercapto-4-methyl-4H-1,2,4-triazole

purum, ≥97.0% (CHN)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5N3S
CAS Number:
Molecular Weight:
115.16
Beilstein:
110580
EC Number:
MDL number:
UNSPSC Code:
12352100

grade

purum

Assay

≥97.0% (CHN)

mp

165-169 °C (lit.)
165-169 °C

SMILES string

Cn1cnnc1S

InChI

1S/C3H5N3S/c1-6-2-4-5-3(6)7/h2H,1H3,(H,5,7)

InChI key

AGWWTUWTOBEQFE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service