Skip to Content
MilliporeSigma
All Photos(1)

Documents

63575

Sigma-Aldrich

D-Mannosamine hydrochloride

≥98.0% (TLC)

Synonym(s):

2-Amino-2-deoxy-D-mannose hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO5 · HCl
CAS Number:
Molecular Weight:
215.63
Beilstein:
3914860
EC Number:
MDL number:
UNSPSC Code:
12352201

Assay

≥98.0% (TLC)

optical activity

[α]20/D −3.7±0.5°, c = 2% in H2O

mp

168 °C (dec.) (lit.)
170-175 °C (dec.)

solubility

H2O: 0.1 g/mL, clear, colorless

SMILES string

Cl[H].N[C@@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O

InChI

1S/C6H13NO5.ClH/c7-3-5(10)4(9)2(1-8)12-6(3)11;/h2-6,8-11H,1,7H2;1H/t2-,3+,4-,5-,6?;/m1./s1

InChI key

QKPLRMLTKYXDST-OHXGPSCHSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jacob Vaya et al.
Vaccine, 27(49), 6869-6876 (2009-09-22)
A variety of protein-coating procedures are used to modify proteins' properties. The principle coating agent used is PEGylation, in which proteins are coated by conjunction to polyethylene glycol (PEG). In the present study, we describe a novel approach that makes
Irma Jiménez et al.
Archives of andrology, 52(6), 455-462 (2006-10-20)
Carbohydrate residues on membrane proteins from sperm are important in gamete interaction. In recent years, Arylsulfatase A (AS-A) has been acquiring an important role from the various putative gamete interaction responsibles in sperm. The aim of this study was to
Tal Gefen et al.
Vaccine, 28(51), 8197-8202 (2010-09-30)
Passive immunization with cross-species antibodies triggers the patient's immune response, thereby preventing repeated treatment. Mannosamine-biotin adduct (MBA) has been described as a masking agent for immunogenic reduction and here, the immunogenicity and biological activity of MBA-coated horse anti-viper venom (hsIgG)
Christopher Brigham et al.
Journal of bacteriology, 191(11), 3629-3638 (2009-03-24)
We characterized the nanLET operon in Bacteroides fragilis, whose products are required for the utilization of the sialic acid N-acetyl neuraminic acid (NANA) as a carbon and energy source. The first gene of the operon is nanL, which codes for
Amanda L Tattersall et al.
Pflugers Archiv : European journal of physiology, 456(3), 501-506 (2008-01-22)
Previous work has shown that interleukin 1 (IL-1) increases the activity of acid extruders in articular chondrocytes, while the H+-adenosine triphosphatase (ATPase) inhibitor bafilomycin can prevent aggrecanase-mediated cartilage degradation. The H+ transport induced by IL-1 may therefore be required for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service