Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

60845

Millipore

Cobalt(II,III) oxide

puriss. p.a., for the determination of sulfur, ≥71% Co basis (KT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
~Co3O4
CAS Number:
Molecular Weight:
240.80
EC Number:
MDL number:
UNSPSC Code:
26111700

grade

puriss. p.a.

Assay

≥71% Co basis (KT)

quality

for the determination of sulfur

impurities

≤0.001% total of sulfur (S)

mp

895 °C (dec.) (lit.)

density

6.11 g/mL at 25 °C (lit.)

cation traces

Ca: ≤200 mg/kg
Cd: ≤10 mg/kg
Cr: ≤10 mg/kg
Cu: ≤10 mg/kg
Fe: ≤100 mg/kg
K: ≤50 mg/kg
Mg: ≤10 mg/kg
Mn: ≤200 mg/kg
Na: ≤200 mg/kg
Ni: ≤1000 mg/kg
Pb: ≤10 mg/kg
Zn: ≤100 mg/kg

SMILES string

O=[Co].O=[Co]O[Co]=O

InChI

1S/3Co.4O

InChI key

LBFUKZWYPLNNJC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Long Ren et al.
Scientific reports, 5, 14229-14229 (2015-09-19)
New and novel 3D hierarchical porous graphene aerogels (HPGA) with uniform and tunable meso-pores (e.g., 21 and 53 nm) on graphene nanosheets (GNS) were prepared by a hydrothermal self-assembly process and an in-situ carbothermal reaction. The size and distribution of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service