Skip to Content
MilliporeSigma
All Photos(1)

Documents

57236

Sigma-Aldrich

Methyl indole-4-carboxylate

purum, ≥98.0% (GC)

Synonym(s):

4-Methoxycarbonylindole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9NO2
CAS Number:
Molecular Weight:
175.18
Beilstein:
141826
MDL number:
UNSPSC Code:
12352100

grade

purum

Assay

≥98.0% (GC)

mp

68-71 °C (lit.)

SMILES string

COC(=O)c1cccc2[nH]ccc12

InChI

1S/C10H9NO2/c1-13-10(12)8-3-2-4-9-7(8)5-6-11-9/h2-6,11H,1H3

InChI key

WEAXQUBYRSEBJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Reactant for preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators
  • Reactant for preparation of cytotoxic agents against multidrug-resistant cancer cells
  • Reactant for preparation of inhibitor for β-tryptase
  • Reactant for preparation of histamine H3 antagonists
  • Reactant for preparation of JNK3 MAP kinase inhibitors
  • Reactant for preparation of nucleosides

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service