Skip to Content
MilliporeSigma
All Photos(1)

Documents

53047

Sigma-Aldrich

5-Carboxy-X-rhodamine triethylamine salt

for fluorescence, ≥90% (HPLC)

Synonym(s):

5-ROX

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H30N2O5 · C6H15N
CAS Number:
Molecular Weight:
635.79
MDL number:
UNSPSC Code:
12352200

grade

for fluorescence

Assay

≥90% (HPLC)

solubility

DMSO: soluble
methanol: soluble

fluorescence

λex 575 nm; λem 600 nm in 0.1 M Tris pH 8.0

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service