Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

42871

Supelco

Diphenylcarbazone, mixture with diphenylcarbazide

for spectrophotometric det. of Cr, Hg

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21

form

powder

Quality Level

quality

for spectrophotometric det. of Cr, Hg

composition

diphenylcarbazide, ~60% HPLC
diphenylcarbazone, ~40% HPLC

concentration

~60%

technique(s)

UV/Vis spectroscopy: suitable

mp

153-157 °C

Other Notes

Reagent for the spectrophotometric det. of various metals

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas R. Crompton
Determination of Anions: A Guide for the Analytical Chemist, 435-435 (1996)
K.L. Cheng et al. et al.
Handbook of Organic Analytical Reagents null
K O Burkey et al.
Biochemistry, 20(10), 2961-2967 (1981-05-12)
The net charge on the photosystem I core complex of Shiozawa et al. (Shiozawa, J. A., Alberte, R. S., & Thornber, J. P. (1974) Arch. Biochem. Biophys. 165, 388-397) has been altered by using a water-soluble carbodiimide to form an
Katsuhiko Yokoi
Biological trace element research, 85(1), 87-94 (2002-03-08)
A colorimetic method is outlined for the determination of the chloride ion in biological samples (blood serum, plasma, and urine). The present method is based on the quantitative reduction of free mercuric ions by chloride ions. Chloride ions form an
Jing Fan et al.
Journal of hazardous materials, 150(2), 343-350 (2007-06-15)
A new solid-phase extractor, diphenylcarbazone-functionalized silica gel has been synthesized and confirmed by IR and Raman spectrometry. The new solid-phase extractor is found to be stable in 1-6 mol L(-1) HCl or H(2)SO(4), and also in common organic solvents. It

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service