Skip to Content
MilliporeSigma
All Photos(1)

Documents

36902

Supelco

PCB No 101 solution

10 μg/mL in isooctane, analytical standard

Synonym(s):

2,2′,4,5,5′-Pentachlorobiphenyl solution, 2,2′,4,5,5′-PCB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H5Cl5
CAS Number:
Molecular Weight:
326.43
Beilstein:
2507418
Ballschmiter Number:
101
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

10 μg/mL in isooctane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

Clc1ccc(Cl)c(c1)-c2cc(Cl)c(Cl)cc2Cl

InChI

1S/C12H5Cl5/c13-6-1-2-9(14)7(3-6)8-4-11(16)12(17)5-10(8)15/h1-5H

InChI key

LAHWLEDBADHJGA-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph E McGraw et al.
Biochemical and biophysical research communications, 344(1), 129-133 (2006-04-18)
Polychlorinated biphenyl IUPAC# 101-PCB 101 (chlorination pattern-2,2',4',5,5') is a common, persistent non-coplanar PCB congener found in the ambient environment but information related to its metabolism in humans is lacking. Previous studies indicate PCB 101 is rapidly metabolized in mammals through
Britt-Marie Bäcklin et al.
Toxicological sciences : an official journal of the Society of Toxicology, 75(1), 154-160 (2003-06-14)
Gray seal females living in the Baltic Sea have been found to exhibit a high prevalence of uterine leiomyomas. These animals are also known to accumulate lipid-soluble PCBs in their blubber. PCBs have documented endocrine-disrupting effects; to investigate whether the
Yoshihisa Kato et al.
Toxicology and applied pharmacology, 263(3), 323-329 (2012-07-18)
The relationships between the changes in the levels of serum total thyroxine (T(4)), serum T(4)-transthyretin (TTR) complex, and accumulation of T(4) in tissues by 2,2',4,5,5'-pentachlorobiphenyl (PentaCB) were examined using wild-type C57BL/6 (WT) and its TTR-deficient (TTR-null) mice. The constitutive level
Paola Pocar et al.
Toxicological sciences : an official journal of the Society of Toxicology, 126(1), 213-226 (2011-12-14)
Several studies indicate that in utero and perinatal exposure to polychlorinated biphenyls (PCBs) induces adverse reproductive effects, but it remains unclear whether such effects may be transmitted to subsequent generations. We therefore investigated the association between maternal exposure to PCBs
Liesbeth Weijs et al.
Toxicology and applied pharmacology, 256(2), 136-145 (2011-08-20)
In the last decade, physiologically based pharmacokinetic (PBPK) models have increasingly been developed to explain the kinetics of environmental pollutants in wildlife. For marine mammals specifically, these models provide a new, non-destructive tool that enables the integration of biomonitoring activities

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service