Skip to Content
MilliporeSigma
All Photos(1)

Documents

36532

Supelco

Fenpiclonil

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H6Cl2N2
CAS Number:
Molecular Weight:
237.08
Beilstein:
5433281
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Clc1cccc(c1Cl)-c2c[nH]cc2C#N

InChI

1S/C11H6Cl2N2/c12-10-3-1-2-8(11(10)13)9-6-15-5-7(9)4-14/h1-3,5-6,15H

InChI key

FKLFBQCQQYDUAM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fenpiclonil is a phenylpyrrole fungicide and a structural analogue of the antibiotic pyrrolnitrin found to be toxic to representatives of Ascomycetes, Basidiomycetes, and Deuteromycetes.

Application

Fenpiclonil may be used as an analytical reference standard for the quantification of the analyte in fruits/vegetables and plant samples using different chromatographic techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Inhalation

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biochemical effects of the phenylpyrrole fungicide fenpiclonil in Fusarium sulphureum (Schlecht)
Jespers.K.BA, et al.
Pesticide Biochemistry and Physiology, 45(2), 116-129 (1993)
Determination of Pesticide Residue Present in Cumin Plant (Nigella orientalis L.) with LC-MS/MS and GC-MS
Ersoy N, et al.
Asian Journal of Chemistry, 28(5), 1011-1011 (2016)
Determination of multiple pesticides in fruits and vegetables using a modified quick, easy, cheap, effective, rugged and safe method with magnetic nanoparticles and gas chromatography tandem mass spectrometry
Li F-Y, et al.
Journal of Chromatography A, 1361, 77-87 (2014)
Similarities in the antifungal activities of fenpiclonil, iprodione and tolclofos-methyl against Botrytis cinerea and Fusarium nivale
Leroux P, et al.
Pestic. Sci., 36(3), 255-261 (1992)
Nouha Bakaraki Turan et al.
Rapid communications in mass spectrometry : RCM, 35(1), e8943-e8943 (2020-09-10)
Pesticides are a group of micropollutants that persist for a long time in the environment and pose threats to life. Much effort has been devoted to developing pre-concentration methods capable of producing samples suitable for the detection of pesticides. However

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service