Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

32962

Supelco

3-Acetyldeoxynivalenol-13C17 solution

25 μg/mL in acetonitrile, analytical standard

Synonym(s):

3-Acetyl-DON 13C labeled

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C17H22O7
CAS Number:
Molecular Weight:
355.23
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

25 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

mass shift

M+17

storage temp.

−20°C

SMILES string

[13CH3][13C](=O)O[13C@@H]1[13CH2][13C@@]2([13CH3])[13C@]3([13CH2]O3)[13C@@H]1O[13C@@H]4[13CH]=[13C]([13CH3])[13C](=O)[13C@@H](O)[13C@]24[13CH2]O

InChI

1S/C17H22O7/c1-8-4-11-16(6-18,13(21)12(8)20)15(3)5-10(23-9(2)19)14(24-11)17(15)7-22-17/h4,10-11,13-14,18,21H,5-7H2,1-3H3/t10-,11-,13-,14-,15-,16-,17+/m1/s1/i1+1,2+1,3+1,4+1,5+1,6+1,7+1,8+1,9+1,10+1,11+1,12+1,13+1,14+1,15+1,16+1,17+1

InChI key

ADFIQZBYNGPCGY-CJTIMMQGSA-N

General description

Certan Vial

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

related product

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2.0 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service