Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

32719

Supelco

Quinoclamine

PESTANAL®, analytical standard

Synonym(s):

2-Amino-3-chloro-1,4-naphthoquinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6ClNO2
CAS Number:
Molecular Weight:
207.61
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

NC1=C(Cl)C(=O)c2ccccc2C1=O

InChI

1S/C10H6ClNO2/c11-7-8(12)10(14)6-4-2-1-3-5(6)9(7)13/h1-4H,12H2

InChI key

OBLNWSCLAYSJJR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Quinoclamine belongs to the class of naphthoquinone compounds.

Application

Quinoclamine may be used as an analytical reference standard for the determination of the analyte in:
  • Honeybees by dispersive solid-phase extraction (dSPE), followed by liquid and gas chromatography coupled with tandem mass spectrometry (LC-MS/MS and GC-MS/MS).
  • Chlorophyll-containing matrix by dSPE and GC-MS/MS.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Sens. 1A - STOT RE 2

Target Organs

Blood,Kidney

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Kwon et al.
Research communications in molecular pathology and pharmacology, 97(2), 215-227 (1997-08-01)
Naphthoquinone compounds have various pharmacological effects such as antiviral, antifungal and anticancer activities. We demonstrated the differentiation of the inducing effect of a naphthoquinone derivative, 2-chloro-3-amino-1,4-nahpthoquinone (NQCA) on the human leukemia cell line U-937. When U-937 cells were treated with
Lijie Hou et al.
Dalton transactions (Cambridge, England : 2003), 48(25), 9234-9242 (2019-06-05)
Green and renewable organic redox molecules are greatly advantageous over conventional inorganic intercalation electrode materials in terms of electrochemical reversibility and cycling stability. However, their electrical insulation prevents them from being used alone as electrode materials for supercapacitors. Herein, 2-amino-3-chloro-1,4-naphthoquinone
Two-step dispersive-solid phase extraction strategy for pesticide multiresidue analysis in a chlorophyll-containing matrix by gas chromatography-tandem mass spectrometry.
Walorczyk S, et al.
Journal of Chromatography A, 1412(2), 22-32 (2015)
Application of bioactivity database of Chinese herbal medicine on the therapeutic prediction, drug development, and safety evaluation.
Cheng HM, et al.
Journal of Ethnopharmacology, 132(2), 429-437 (2010)
Multi-residue method for the determination of pesticides and pesticide metabolites in honeybees by liquid and gas chromatography coupled with tandem mass spectrometry-Honeybee poisoning incidents.
Kiljanek T, et al.
Journal of Chromatography A, 1435(2), 100-114 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service