Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31481-M

Sigma-Aldrich

Sodium sulfate

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., anhydrous, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2SO4
CAS Number:
Molecular Weight:
142.04
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent
puriss. p.a.

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

vapor pressure

<0.1 hPa ( 20 °C)

Assay

≥99.0%

form

crystalline

impurities

≤0.0005% heavy metals (by ICP-OES)
≤0.01% in water insoluble matters
≤0.03% free alkali (as NaOH)
≤0.05% free acid (as H2SO4)
≤5 ppm total nitrogen (N)

loss

≤0.5% loss on ignition, 800 °C

pH

5.2-8.0 (20 °C, 5%)

mp

884 °C (lit.)

density

2.68 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤10 ppm
phosphate (PO43-): ≤0.001%

cation traces

As: ≤0.0001%
Ca: ≤20 ppm
Fe: ≤5 ppm
K: ≤0.01%
Mg: ≤25 ppm

suitability

complies for appearance of solution

SMILES string

[Na+].[Na+].[O-]S([O-])(=O)=O

InChI

1S/2Na.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

PMZURENOXWZQFD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

NaSO4 can also be used as a drying agent in organic synthesis for the removal of traces of water from organic extracts.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service