Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

301310

Sigma-Aldrich

2-Ethyl-2-thiopseudourea hydrobromide

98%

Synonym(s):

S-Ethylisothiourea hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5SC(=NH)NH2 · HBr
CAS Number:
Molecular Weight:
185.09
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

Assay

98%

form

powder or crystals

mp

88-90 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Br.CCSC(N)=N

InChI

1S/C3H8N2S.BrH/c1-2-6-3(4)5;/h2H2,1H3,(H3,4,5);1H

InChI key

SWXXKWPYNMZFTE-UHFFFAOYSA-N

Application

Reagent used in the synthesis of pyrimidines.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claus Martin et al.
Cardiovascular research, 74(1), 46-55 (2007-01-30)
Nitric oxide (NO) synthesis by NO synthases (NOS) requires oxygen. However, although counterintuitive, NO synthesis is increased in ischemic myocardium. Accordingly, mechanisms independent of the NOS pathway have been suggested to contribute to NO synthesis during ischemia. NO initiates detrimental
A L Bardell et al.
The Journal of pharmacology and experimental therapeutics, 296(2), 252-259 (2001-02-13)
Experiments were performed to investigate whether nitric-oxide synthase (NOS) activity can be detected in vascular smooth muscle (VSM) from 12- to 14-week streptozotocin (STZ)-diabetic rats. Concentration-response curves to norepinephrine (NE) of superior mesenteric arteries from diabetic and age- and gender-matched
Barbara T Alexander et al.
Hypertension (Dallas, Tex. : 1979), 39(2 Pt 2), 586-590 (2002-03-08)
Acute, nonselective nitric oxide synthase inhibition in the pregnant rat decreases glomerular filtration rate and renal plasma flow, suggesting a role for nitric oxide in mediating renal vasodilation during pregnancy. As mid-gestation in the rat is associated with a significant
E Alhan et al.
The European journal of surgery = Acta chirurgica, 164(9), 697-702 (1998-09-05)
To investigate the effects of the constitutive nitric oxide (NO) synthase inhibitor NG-nitro-L-arginine methyl ester (L-Name) and the inducible NO synthase inhibitor amino ethyl-isothiourea (AE-ITU) on acute necrotising pancreatitis (ANP) in rats. Laboratory study. Medical school, Turkey. Morbidity, mortality, effects
W K Alderton et al.
The Biochemical journal, 332 ( Pt 1), 195-201 (1998-05-13)
Nitric oxide synthases (NOS) have a bidomain structure comprised of an N-terminal oxygenase domain and a C-terminal reductase domain. The oxygenase domain binds haem, (6R)-5,6,7,8-tetrahydro-l-biopterin (tetrahydrobiopterin) and arginine, is the site where nitric oxide synthesis takes place and contains determinants

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service