Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24218

Sigma-Aldrich

Cyclohexanone

puriss., ≥99.5% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H10(=O)
CAS Number:
Molecular Weight:
98.14
Beilstein:
385735
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

3.4 (vs air)

vapor pressure

3.4 mmHg ( 20 °C)

grade

puriss.

Assay

≥99.5% (GC)

autoignition temp.

788 °F

expl. lim.

1.1 %, 100 °F
9.4 %

impurities

≤0.2% water (Karl Fischer)

refractive index

n20/D 1.450 (lit.)

bp

155 °C (lit.)

mp

−47 °C (lit.)

density

0.947 g/mL at 25 °C (lit.)

SMILES string

O=C1CCCCC1

InChI

1S/C6H10O/c7-6-4-2-1-3-5-6/h1-5H2

InChI key

JHIVVAPYMSGYDF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aldol Condensation of Cyclohexanone and Synthesis of Acetal or Ketals [J]. J. Jilin
Yiezhi L, et al.
Journal of Jilin University (Science Edition) / Chi Lin Ta Hsueh Hsueh Pao (Li Hsueh Pan ), 2, 021-021 (1989)
194. A synthesis of vitamin a from cyclohexanone.
Attenburrow J, et al.
Journal of the Chemical Society, 1094-1111 (1952)
Huizhen Liu et al.
Science (New York, N.Y.), 326(5957), 1250-1252 (2009-12-08)
Cyclohexanone is an industrially important intermediate in the synthesis of materials such as nylon, but preparing it efficiently through direct hydrogenation of phenol is hindered by over-reduction to cyclohexanol. Here we report that a previously unappreciated combination of two common
Saulo F P Braga et al.
European journal of medicinal chemistry, 71, 282-289 (2013-12-11)
A series of bis-(arylmethylidene)-cycloalkanones was synthesized by cross-aldol condensation. The activity of the compounds was evaluated against amastigotes forms of Trypanosoma cruzi and promastigotes forms of Leishmania amazonensis. The cytotoxicity of the active compounds on uninfected fibroblasts or macrophages was
Viktória Fábos et al.
Chemistry, an Asian journal, 7(11), 2638-2643 (2012-09-07)
The processing of renewable feedstocks to platform chemicals and, to a lesser degree, fuels is a key part of sustainable development. In particular, the combination of lignocellulosic biomass with hydrothermal upgrading (HTU), using high temperature and pressure water (HTPW), is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service