Skip to Content
MilliporeSigma
All Photos(1)

Documents

11686

Supelco

S,S,S-Tris(2-ethylhexyl)phosphorotrithioate

Selectophore

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H51OPS3
CAS Number:
Molecular Weight:
482.83
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

product line

Selectophore

SMILES string

CCCCC(CC)CSP(=O)(SCC(CC)CCCC)SCC(CC)CCCC

InChI

1S/C24H51OPS3/c1-7-13-16-22(10-4)19-27-26(25,28-20-23(11-5)17-14-8-2)29-21-24(12-6)18-15-9-3/h22-24H,7-21H2,1-6H3

InChI key

ZYMYBLYKQDZTEH-UHFFFAOYSA-N

General description

S,S,S-Tris(2-ethylhexyl)phosphorotrithioate is a solvent mediating reagent. It is a phosphorus compound containing phosphoryl group. It has high selectivity towards inorganic cations.

Application

It may be used as solvent mediator in constructing mexiletine-sensitive membrane electrode and serotonin-selective membrane electrode.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takashi Katsu et al.
Analytical and bioanalytical chemistry, 387(6), 2057-2064 (2007-02-01)
S,S,S-tris(2-ethylhexyl) phosphorotrithioate proved to be an effective solvent mediator for constructing a mexiletine-sensitive membrane electrode in combination with an ion-exchanger, sodium tetrakis[3,5-bis(2-methoxyhexafluoro-2-propyl)phenyl]borate. Among a series of phosphorus compounds containing phosphoryl (P=O) groups, this solvent mediator showed the highest sensitivity to
Serotonin-Selective Membrane Electrode Made with the Solvent Mediator S,S,S-Tris(2-ethylhexyl)phosphorotrithioate
T.Katsu, H.Hirodo
Sensor Letters, 1, 99-101 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service