Skip to Content
MilliporeSigma
All Photos(1)

Documents

04619

Supelco

2-{2-[7-(Diethylamino)-2-oxo-2H-1-benzopyran-3-yl]ethenyl}-1,3,3-trimethyl-3H-indolium iodide

for fluorescence

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H29IN2O2
CAS Number:
Molecular Weight:
528.43
Beilstein:
22103985
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21

grade

for fluorescence

Quality Level

Assay

≥98.0% (HPLC)

technique(s)

UV/Vis spectroscopy: suitable

solubility

methanol: 1 mg/mL, clear (violet to deep violet)

fluorescence

λex 406 nm; λem 542.0 nm±10 nm (in the presence of cyanide)

SMILES string

C[N+]1=C(/C=C/C2=CC3=C(C=C(N(CC)CC)C=C3)OC2=O)C(C)(C)C4=CC=CC=C41.[I-]

InChI

1S/C26H29N2O2.HI/c1-6-28(7-2)20-14-12-18-16-19(25(29)30-23(18)17-20)13-15-24-26(3,4)21-10-8-9-11-22(21)27(24)5;/h8-17H,6-7H2,1-5H3;1H/q+1;/p-1

InChI key

OZKWMKLMZONDEZ-UHFFFAOYSA-M

Application

fluorescence dye for ratiometric determination of cyanide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xin Lv et al.
Chemical communications (Cambridge, England), 47(48), 12843-12845 (2011-11-03)
A new ratiometric fluorescent cyanide probe was developed based on the nucleophilic attack of CN(-) toward the indolium group of a hybrid coumarin-hemicyanine dye, by which high selectivity as well as large emission shift could be achieved.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service