Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

05-2690

Sigma-Aldrich

Chloramine T trihydrate

JIS special grade, ≥98.0%

Synonym(s):

N-Chloro-p-toluenesulfonamide sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO2NClNa · 3H2O
CAS Number:
Molecular Weight:
281.69
Beilstein:
3924168
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:

grade

JIS special grade

Assay

≥98.0%

availability

available only in Japan

mp

167-170 °C (lit.)

SMILES string

O.O.O.Cc1ccc(cc1)S(=O)(=O)N([Na])Cl

InChI

1S/C7H7ClNO2S.Na.3H2O/c1-6-2-4-7(5-3-6)12(10,11)9-8;;;;/h2-5H,1H3;;3*1H2/q-1;+1;;;

InChI key

NZYOAGBNMCVQIV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

377.6 °F - closed cup

Flash Point(C)

192 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service