Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

01-0380

Sigma-Aldrich

Acetic anhydride

JIS special grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CO)2O
CAS Number:
Molecular Weight:
102.09
Beilstein:
385737
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

JIS special grade

vapor density

3.5 (vs air)

vapor pressure

10 mmHg ( 36 °C)
4 mmHg ( 20 °C)

form

liquid

autoignition temp.

629 °F

expl. lim.

10.3 %

availability

available only in Japan

refractive index

n20/D 1.390 (lit.)

bp

138-140 °C (lit.)

mp

−73 °C (lit.)

density

1.08 g/mL (lit.)

storage temp.

15-25°C

SMILES string

CC(=O)OC(C)=O

InChI

1S/C4H6O3/c1-3(5)7-4(2)6/h1-2H3

InChI key

WFDIJRYMOXRFFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

120.2 °F

Flash Point(C)

49 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lakshmi Pillai-Kastoori et al.
PLoS genetics, 10(7), e1004491-e1004491 (2014-07-11)
Ocular coloboma is a sight-threatening malformation caused by failure of the choroid fissure to close during morphogenesis of the eye, and is frequently associated with additional anomalies, including microphthalmia and cataracts. Although Hedgehog signaling is known to play a critical
Maciej Podgórski
Dental materials : official publication of the Academy of Dental Materials, 27(8), 748-754 (2011-04-26)
In this study acetyloxypropylene dimethacrylate was synthesized and proposed as an alternative monomer for triethyleneglycol dimethacrylate (TEGDMA) in dental mixtures. The monomer was prepared by the reaction of glycerol dimethacrylate with acetic anhydride. The exchange reaction was carried out in
David G Simmons et al.
BMC genomics, 9, 352-352 (2008-07-30)
The Prolactin (PRL) hormone gene family shows considerable variation among placental mammals. Whereas there is a single PRL gene in humans that is expressed by the pituitary, there are an additional 22 genes in mice including the placental lactogens (PL)
M Raro et al.
Journal of mass spectrometry : JMS, 49(6), 509-521 (2014-06-11)
The detection of anabolic androgenic steroids (AAS) is one of the most important topics in doping control analysis. Gas chromatography coupled to (tandem) mass spectrometry (GC-MS(/MS)) with electron ionization and liquid chromatography coupled to tandem mass spectrometry have been traditionally
Heidi I Chen et al.
Development (Cambridge, England), 141(23), 4500-4512 (2014-11-08)
Identifying coronary artery progenitors and their developmental pathways could inspire novel regenerative treatments for heart disease. Multiple sources of coronary vessels have been proposed, including the sinus venosus (SV), endocardium and proepicardium, but their relative contributions to the coronary circulation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service