Skip to Content

We are planning system maintenance on January 25, 2025, from 1:00 PM to 4:00 PM CST. This will impact both web and offline transactions, including online orders, quotes, price and availability checks, and order status inquiries. We apologize for any inconvenience.

MilliporeSigma
All Photos(1)

Key Documents

RIE156324985

Rieke Metals, Inc.

2,4-dimethyl-3'-thiomorpholinomethyl benzophenone

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
325.47
MDL number:

Pricing and availability is not currently available.

Iupac Name

(2,4-dimethylphenyl)[3-(4-thiomorpholinylmethyl)phenyl]methanone

Inchi Code

1S/C20H23NOS/c1-15-6-7-19(16(2)12-15)20(22)18-5-3-4-17(13-18)14-21-8-10-23-11-9-21/h3-7,12-13H,8-11,14H2,1-2H3

InChI key

YHMXOZAULKTQLE-UHFFFAOYSA-N

Purity

97%

Country of Origin

US

Product Link

https://riekemetals.com/products-services/catnum/6125-0700-27

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service