Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ST1507

Sigma-Aldrich

Anti-AKR1B10 Mouse mAb (1A6)

liquid, clone 1A6, Calbiochem®

Synonym(s):

Anti-Aldo-Keto Reductase 1B10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1A6, monoclonal

form

liquid

does not contain

preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG2a

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AKR1B10(57016)

General description

Anti-AKR1B10. Mpuse monoclonal, clone 1A6 recognizes the ~37 kDa AKR1B10 protein in HepG2 and HeLa cells and human colon tissue. It is validated for ELISA, WB, ICC, and paraffin sections.
Purified mouse monoclonal antibody. Recognizes the ~37 kDa AKR1B10 protein.
Recognizes the ~37 kDa AKR1B10 protein in HepG2 and HeLa cells and human colon tissue.

Immunogen

A recombinant polypeptide corresponding to amino acids 76-144 of human AKR1B10, expressed as a GST fusion protein
NA

Application




ELISA (see comments)
Immunoblotting (1-5 g/ml)
Immunocytochemistry (10 g/ml)
Paraffin Sections (3 g/ml)

Warning

Toxicity: Regulatory Review (Z)

Physical form

In PBS, pH 7.2.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
HepG2 cells, Human Colon tissue, HeLa cells

Other Notes

Ahn K.H., et al. 2010 JOURNAL OF HEALTH SCIENCE56, 287.
Satow, R., et al. 2010. Clin. Cancer Res.16, 2518.
For ELISA, this antibody can be used as the capture antibody; recommended concentration is 10 ng/ml, 1 µg/well, 100 µl total volume. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer A Harbottle et al.
Analytical biochemistry, 592, 113583-113583 (2020-01-17)
Improved health span and lifespan extension in a wide phylogenetic range of species is associated with the induction of the environmental cell stress response through a signalling pathway regulated by the transcription factor Nrf2. Phytochemicals which stimulate this response may

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service