Key Documents
RPSTLTVP1
Millipore
Rapid Sterility Testing Validation Protocol
Letter Format, Milliflex Rapid PQ Protocol, for North America only
About This Item
UNSPSC Code:
41116500
NACRES:
JA.11
Pricing and availability is not currently available.
Quality Level
compatibility
for use with Milliflex® Rapid Systems.
General description
Rapid Sterility Testing Validation Protocol is designed as a ready to use step by step guide to perform PQ activities of rapid sterility testing using Milliflex® Rapid system. The PQ protocol will enable the QC/QA Labs to initiate the performance qualification after the IQ andOQ have been completed following the respective separate protocol. The guidelines are GMP compliant and follow EP, USP and JP regulatory guidelines.
Application
Rapid Sterility Testing Validation Protocol is used to validate rapid sterility testing procedures with ease and consistent performance while following regulatory guidelines.
Legal Information
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany