Skip to Content
MilliporeSigma
All Photos(1)

Documents

MABX36

Sigma-Aldrich

Anti-Norovirus genogroup II

Synonym(s):

Anti-Norovirus Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

clone

monoclonal

form

solution

species reactivity

all

packaging

pkg of 1 mg (MABX36-1KC)
pkg of 10 mg (MABX36-10KC)

technique(s)

ELISA: suitable
immunodiffusion: suitable

isotype

IgG1

relevant disease(s)

infectious diseases

shipped in

wet ice

storage temp.

2-8°C

Specificity

Norovirus GII

Immunogen

Norovirus GII.4 VLP

Application

This mouse monoclonal antibody, Cat. No. MABX36, is tested for use in Lateral Flow Assay and ELISA, for the detection of Norovirus GII.

Target description

Noroviruses are a genetically diverse positive stranded RNA virus of the Caliciviridae family. Genotypes I, II and IV can cause the sudden onset of sever vomiting and diarrhea.  The virus is highly contagious and commonly spread through contaminated food, water and through close contact with infected persons.

Linkage

Recommended Matched Pair MABX37 capture and MABX36 detection

Physical form

Purified mouse monoclonal in buffer containing 10 mM Phosphate Buffered Saline, pH 7.0 with 0.09% Sodium Azide

Reconstitution

Please refer to Certificate of Analysis for concentration.

Storage and Stability

Store at 2-8 °C. For long-term storage, freeze at -20 °C

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service