Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABT1533

Sigma-Aldrich

Anti-Actin Antibody, clone 5-16 H1C8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

antibody form

purified antibody

clone

MAb GEa (5-16 H1C8)
monoclonal

purified by

(Ligatrap)

species reactivity

mouse, human

concentration

(Please refer to lot specific datasheet.)

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgM

UniProt accession no.

General description

Purified actin (ACT1) from Arabidopsis thaliana plant.

Specificity

Clone 5-16 H1C8 is a mouse monoclonal antibody that detects Actin in a wide range of species.

Application

ELISA Analysis: A representative lot detected Actin in ELISA applications (Kandasamy, M.K., et. al. (1999). Plant J. 18(6):681-91).

Western Blotting Analysis: A representative lot detected Actin in Western Blotting applications (Kandasamy, M.K., et. al. (2012). Plant Cell. 24(5):2041-57; Kandasamy, M.K., et. al. (1999). Plant J. 18(6):681-91).

Quality

Evaluated by Western Blotting in human skeletal muscle tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Actin in human skeletal muscle tissue lysate.

Physical properties

~44 kDa observed; 41.86 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Purified mouse monoclonal antibody IgM in PBS without azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Other Notes

Human. Predicted to react with all eukaryotes based on 100% sequence homology.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service