Skip to Content
MilliporeSigma
All Photos(1)

Documents

MABS1816

Sigma-Aldrich

Anti-LRIG1 Antibody, clone 2D1.1

clone 2D1.1, from mouse

Synonym(s):

Leucine-rich repeats and immunoglobulin-like domains protein 1, LIG-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2D1.1, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... LRIG1(26018)

General description

Leucine-rich repeats and immunoglobulin-like domains protein 1 (UniProt: Q96JA1; also known as LIG-1) is encoded by the LRIG1 (also known as LIG1) gene (Gene ID: 26018) in human. LIG-1 is a widely expressed single-pass type I membrane protein that acts as a feedback negative regulator of signaling by receptor tyrosine kinases, through a mechanism that involves enhancement of receptor ubiquitination and accelerated intracellular degradation. LIG-1 contains 3 Ig-like C2-type (immunoglobulin-like) domains, 15 LRR (leucine-rich) repeats, 1 1 LRRCT domain, and 1LRRNT domain. The LRRs and the Ig-domains are each sufficient for EGFR/ERBB1 binding and this interaction is abolished only when these two domains are deleted. LIG-1 is highly expressed by intestinal stem cells and controls the size of the intestinal stem-cell niche by regulating the amplitude of growth-factor signaling. It is also reported to control stem-cell proliferation in epidermis. LIG-1 is reported to be expressed in a gradient with highest levels in intestinal stem cells and it is absent from Paneth cells. About one-third of all cells in the intestinal crypts are shown to express LIG-1. (Ref.: Wong VWY., et al. (2012). Nat. Cell Biol. 14(4); 401-408).

Specificity

Clone 2D1.1 detects Leucine-rich repeats and immunoglobulin-like domains protein 1 (LIG-1) in human. It targets an epitope within 134 amino acids from the C-terminal end.

Immunogen

GST/His-tagged recombinant fragment corresponding to 134 amino acids from the C-terminal end of human Leucine-rich repeats and immunoglobulin-like domains protein 1 (LRIG1).

Application

Anti-LRIG1 Antibody, clone 2D1.1, Cat. No. MABS1816, is a mouse monoclonal antibody that targets human Leucine-rich repeats and immunoglobulin-like domains protein 1 (LRIG1) and has been tested for use in Western Blotting.
Research Category
Signaling

Quality

Evaluated by Western Blotting in LNCaP cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected LRIG1 in 10 µg of LNCaP cell lysate.

Target description

~150 kDa observed; 119.11 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service