Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABS1272

Sigma-Aldrich

Anti-Endothelial Protein C Receptor Antibody, clone 1560

clone 1560, from rat

Synonym(s):

Activated protein C receptor, APC receptor, Centrocyclin, Centrosomal protein CCD41, Endothelial cell protein C receptor, CD201

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1560, monoclonal

species reactivity

mouse

technique(s)

flow cytometry: suitable
inhibition assay: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... Procr(19124)

General description

Endothelial protein C receptor (UniProt: Q64695; also known as EPCR, Activated protein C receptor, APC receptor, Centrocyclin, Centrosomal protein CCD41, Endothelial cell protein C receptor, CD201) is encoded by the Procr (also known as Epcr) gene (Gene ID: 19124) in murine species. EPCR is a multi-functional single-pass type I membrane protein that is expressed in endothelial cells. Its expression has also been reported in hematopoietic, neuronal, epithelial progenitor cells, and breast cancer stem cells. It binds to activated protein C and enhances protein C activation by the thrombin-thrombomodulin complex and thereby plays a role in the protein C pathway controlling blood coagulation. It is also shown to play a role in mediating activated protein C-induced cytoprotective effects, including antiapoptotic, anti-inflammatory, and barrier stabilization. EPCR exhibits sequence and 3-dimensional structural homology with the major histocompatibility class 1/CD1 family of proteins, particularly CD1d. It contains a tightly bound phospholipid in the antigen presenting groove and phosphatidylcholine is shown to be the major phospholipid bound to human EPCR. Removal of this lipid results in the loss of protein C binding.

Specificity

Clone 1560 specifically detects endothelial protein C receptor in murine species.

Immunogen

A full length recombinant mouse endothelial protein C receptor.

Application

Anti-Endothelial Protein C Receptor, clone 1560, Cat. No. MABS1272, is a rat monoclonal antibody for detection of murine Endothelial Protein C Receptor and is tested for use in Flow Cytometry and Inhibition Assays.
Flow Cytometry Analysis: A representative lot detected Endothelial Protein C Receptor in Flow Cytometry applications (Li, W., et. al. (2005). J Thromb Haemost. 3(7):1351-9; Balazs, A.B., et. al. (2006). Blood. 107(6):2317-21).

Inhibition Analysis: A representative lot detected Endothelial Protein C Receptor in Inhibition applications (Li, W., et. al. (2005). J Thromb Haemost. 3(7):1351-9; Disse, J., et. al. (2011). J Biol Chem. 286(7):5756-67).
Research Category
Signaling

Quality

Evaluated by Flow Cytometry in bEND.3 mouse endothelial cells.

Flow Cytometry Analysis: 1 µg of this antibody detected Endothelial Protein C Receptor in one million bEND.3 mouse endothelial cells.

Target description

27.19 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified rat monoclonal antibody IgG2b in PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service