Skip to Content
MilliporeSigma
All Photos(1)

Documents

MABE956

Sigma-Aldrich

Anti-NFE2-related factor 1/Nrf1 Antibody, clone 17

clone 17, from rat

Synonym(s):

Nuclear factor erythroid 2-related factor 1, NF-E2-related factor 1, NFE2-related factor 1, Nuclear factor, erythroid derived 2, like 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

17, monoclonal

species reactivity

mouse

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Nfe2(18022)

General description

Nuclear factor erythroid 2-related factor 1 (NFE2-related factor 1/Nrf1), also known as NF-E2-related factor 1, NFE2-related factor 1, Nuclear factor erythroid derived 2 like 1, and encoded by the gene name Nfe2l1 and Nrf1, is a member of the bZIP family and the CNC subfamily. Isoform 3 of Nrf1 interacts with the extended kappa 3 site of the TNF alpha promoter after Fc gamma RIII stimulation and participates in the induction of this cytokine. Studies have suggested that isoform 1 of Nrf1 is either inactive or represses the transcriptional activation. Isoform 2 of Nrf1 has been shown to activate the expression of antioxidant response element (ARE) containing reporters and genes in cells, and may play a role in oxidative stress response. Other studies have suggested that isoform 1 also activates ARE-driven genes. Levels of Nrf1 isoform 2 in white fat, lung and liver are increased compared to isoform 1 (at protein level). Isoform 1 and isoform 2 of Nrf1 are widely expressed including kidney, brown fat, white fat, large intestine, small intestine, stomach, lung, brain and liver. Isoform 2 levels are elevated in brown fat and brain, but are reduced in liver compared to isoform 1 levels. Isoform 1 and isoform 2 are also expressed in mouse embryonic fibroblasts (MEF).

Immunogen

His-tagged recombinant protein corresponding to mouse NFE2-related factor 1/Nrf1.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors
This Anti-NFE2-related factor 1/Nrf1 antibody is validated for use in Western Blotting for the detection of NFE2-related factor 1/Nrf1.
Western Blotting Analysis: A representative lot detected NFE2-related factor 1/Nrf1 in MEF cells stimulated with MG132 (Tsuchiya, Y., et al. (2011) Mole. Cell Biol. 31(22):4500-4512).

Quality

Evaluated by Western Blotting in MG132 treated NIH-3T3 cell lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected NFE2-related factor 1/Nrf1 in 10 µg of MG132 treated NIH-3T3 cell lysate.

Target description

~85 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service