Skip to Content
MilliporeSigma
All Photos(1)

Documents

MAB505B

Sigma-Aldrich

Mouse Anti-Rat IgG2b γ2b chain Antibody, biotin conjugate

Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

mouse

Quality Level

conjugate

biotin conjugate

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

monoclonal

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
flow cytometry: suitable

shipped in

wet ice

target post-translational modification

unmodified

General description

IgG is one of the most abundant protein in human serum. IgG is important for our defence against microorganisms and the molecules are produced by B-lymphocytes as a part of our adaptive immune response. The IgG molecule has two separate functions; to bind to the pathogen that elicited the response and to recruit other cells and molecules to destroy the antigen. The variability of the IgG pool is generated by somatic recombination and the number of specificities in an individual at a given time point is estimated to be 1011 variants.

Specificity

The antibody reacts with the γ2b heavy chain (Fc) of rat IgG2b.

Immunogen

Epitope: IgG2b γ2b chain
Prepared from rat IgG2b γ2b chain.

Application

Mouse anti-Rat IgG2b γ2b chain Antibody, biotin conjugate is an antibody against Rat IgG2b γ2b chain for use in ELISA, FC.
Research Category
Secondary & Control Antibodies
Research Sub Category
Fragment Specific Secondary Antibodies

Physical form

Affinity Chromatography
Purified by affinity chromatography. Liquid in 1.0 mL PBS/NaN3.

Storage and Stability

Maintain refrigerated at 2°-8°C under sterile conditions for up to twelve months from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service