Skip to Content
MilliporeSigma
All Photos(1)

Documents

MAB3738

Sigma-Aldrich

Anti-PML Antibody, clone 36.1-104

ascites fluid, clone 36.1-104, Chemicon®

Synonym(s):

Promyelocytic Leukemia Protein, Tripartite Motif Protein 19

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

ascites fluid

antibody product type

primary antibodies

clone

36.1-104, monoclonal

species reactivity

mouse

should not react with

human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2b

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... PML(5371)

General description

PML protein is a tripartite motif (TRIM)-containing nuclear protein that may function as, among other things, a transcription factor, a coactivator of nuclear receptors (Ruggerio, 2000), a regulator of apoptosis (Guo, 2000), a mediator of interferon-induced antiviral response (Regand and Chelbi-Aliz, 2001), and as a suppressor of growth and oncogenic transformation (Mu, 1997). PML is localized to the nucleoplasm and in distinct subnuclear structures referred to as Promyelocytic Leukemia Bodies (also known as Nuclear Domain 10). Localization of PML to Promyelocytic Leukemia Bodies requires modification of PML protein by the Small Ubiquitin Modifier (SUMO) and is required for proper formation and integrity of these subnuclear structures. At least 14 splice variants of PML ranging in molecular weight from 48-97 kDa (predicted) have been described in the literature. The functional significance of the various splice variants is not well understood. In patients with Acute Promyelocytic Leukemia, the PML gene is involved in at least two specific chromosomal translocations that result in the expression of chimeric proteins with the Retinoic Acid Receptor alpha (RARalpha DNA- and Hormone-binding domains; Pandolfi, 2001). All isoforms of PML, as well as the PML-RARalpha chimeric proteins expressed in Type A and Type B APL contain an identical N-terminus but vary in the C-terminal portion of the protein (Jenson, 2001).

Specificity

Recognizes mouse PML (Promyelocytic Leukemia protein). On western blots of protein extracts from mouse embryo fibroblast (MEF1 cells), MAB3738 recognizes a band migrating at approximately 106 kDa corresponding to PML.

Immunogen

His-tagged PML fusion protein corresponding to amino acids 1-581 of mouse PML.

Application

Anti-PML Antibody, clone 36.1-104 is a Mouse Monoclonal Antibody for detection of PML also known as Promyelocytic Leukemia Protein & has been validated in ICC, IP & WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
Western blot (1:500)

Immunoprecipitation

Immunocytochemistry (1:100)

Optimal working dilutions must be determined by end user.

Target description

106 kDa

Physical form

Ascites fluid containing no preservatives.
Unpurified

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
POSITIVE CONTROL: Mouse embryo fibroblasts (MEF1 cells).

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tracy Dagher et al.
The Journal of experimental medicine, 218(2) (2020-10-20)
Interferon α (IFNα) is used to treat JAK2V617F-driven myeloproliferative neoplasms (MPNs) but rarely clears the disease. We investigated the IFNα mechanism of action focusing on PML, an interferon target and key senescence gene whose targeting by arsenic trioxide (ATO) drives
Marie-Odile Baudement et al.
Genome research, 28(11), 1733-1746 (2018-10-06)
The mammalian cell nucleus contains numerous discrete suborganelles named nuclear bodies. While recruitment of specific genomic regions into these large ribonucleoprotein (RNP) complexes critically contributes to higher-order functional chromatin organization, such regions remain ill-defined. We have developed the high-salt-recovered sequences-sequencing
Arkadiy K Golov et al.
PloS one, 10(10), e0139855-e0139855 (2015-10-06)
The extremely high concentration of macromolecules in a eukaryotic cell nucleus indicates that the nucleoplasm is a crowded macromolecular solution in which large objects tend to gather together due to crowding forces. It has been shown experimentally that crowding forces
Sonia Missiroli et al.
Cell reports, 16(9), 2415-2427 (2016-08-23)
The precise molecular mechanisms that coordinate apoptosis and autophagy in cancer remain to be determined. Here, we provide evidence that the tumor suppressor promyelocytic leukemia protein (PML) controls autophagosome formation at mitochondria-associated membranes (MAMs) and, thus, autophagy induction. Our in vitro and
Srikanth Talluri et al.
PloS one, 8(8), e72236-e72236 (2013-08-13)
The Retinoblastoma protein (pRB) is a key tumor suppressor that is functionally inactivated in most cancers. pRB regulates the cell division cycle and cell cycle exit through protein-protein interactions mediated by its multiple binding interfaces. The LXCXE binding cleft region

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service