Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HG00C134

Millipore

Peptone Water Buffered

pH 7.0±0.2 (25 °C, after autoclaving)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352111

composition

di-Sodium Hydrogen Phosphate, 3.5 g/L
Peptone, 10.0 g/L
Potassium Dihydrogen Phosphate, 1.5 g/L
Sodium Chloride, 5.0 g/L

packaging

pkg of 5 kg (HG00C134.5)
poly bottle of 12 × 500 g (HG00C134.500)

pH

7.0±0.2 (25 °C, after autoclaving)

storage temp.

15-25°C

Application

A liquid medium for the preliminary, non-selective enrichment of bacteria, particularly of pathogenic Enterobacteriaceae, from meat, milk and other foods.

Preparation Note

Suspend 20g in 1 litre demineralised water. Dispense into final containers and autoclave at 121°C for 15 minutes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service