Skip to Content
MilliporeSigma
All Photos(1)

Documents

GF53

Sigma-Aldrich

Anti-Tissue Plasminogen Activator (Ab-1) Mouse mAb (GMA-043)

liquid, clone GMA-043, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

GMA-043, monoclonal

form

liquid

does not contain

preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PLAT(5327)

General description

Purified mouse monoclonal antibody. Recognizes the ~60 kDa tPA protein.
Recognizes the ~60 kDa tPA protein.
This Anti-Tissue Plasminogen Activator (Ab-1) Mouse mAb (GMA-043) is validated for use in ELISA, Immunoblotting for the detection of Tissue Plasminogen Activator (Ab-1).

Immunogen

Human
purified human tissue plasminogen activator (tPA)

Application


ELISA (1-5 g/ml)
Immunoblotting (1-5 g/ml)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 150 mM NaCl, 10 mM sodium phosphate, 1% mannitol, pH 7.4.

Reconstitution

Following initial thaw, aliquot and freeze (-20°).

Analysis Note

Positive Control
Human tPA

Other Notes

Antibody should be titrated for optimal results in individual systems.
Hartleib, J., et al. 2000. Blood96, 2149.
Wiman, B., et al. 2000. Arterioscler Thromb. Vasc. Biol.20, 2019.
Pourtau, J., et al. 1998. Blood Coagul Fibrinolysis9, 609.
Van, D.E., et al. 1995. Blood85, 3510.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service