Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

EZSGR0M02

Millipore

Grommets for EZ-Stream® Vacuum Pump

Hole Protectors for EZ-Stream® Vacuum Pump

Synonym(s):

Grommet Ring, Grommet Seal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116500
NACRES:
NB.79
Pricing and availability is not currently available.

material

EPDM rubber (Grommet Ring)

feature

repair kit

packaging

pkg of 2 units

manufacturer/tradename

EZ-Stream®

application(s)

bioburden testing
food and beverages
industrial qc
pharmaceutical
water monitoring

compatibility

for use with EZ-Stream® Vacuum Pump

General description

Grommet Rings for EZ-Stream® Vacuum Pump are designed to protect the tubing used for liquid transfer from sharp edges in the system. Grommet rings are necessary when hole reinforcements are needed to prevent fraying.
This reference includes two (2) grommets to replace original parts that protect the inlet and outlet hoses barbs connectors and ensure their good positioning.
The EZ-Stream® Vacuum Pump is ideal to be used both on work bench and laminar flow hood, where its small footprint saves laboratory space.

Application

Grommet Rings for EZ-Stream® Vacuum Pump is used to prevent tubing damage that are intended for liquid transfer.

Legal Information

EZ-STREAM is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service