Documents
EZCURVP0W
Millipore
EZ-Pak® External Power Supply
for AC Powered Dispenser
About This Item
UNSPSC Code:
41116129
NACRES:
NB.79
Quality Level
manufacturer/tradename
EZ-Pak®
application(s)
bioburden testing
food and beverages
compatibility
for use with EZ-PAK® Curve
General description
EZ-Pak® External Power Supply is an accessory system designed to enable EZ-Pak® Membrane Dispenser to work without a direct power supply. The Lithium Ion Battery upon fully charged can dispense 10,000 membranes. The EZ-Pak® Curve saves considerable amount of time in opening the membranes for easy use and thereby reduces the risk of contamination.
Application
EZ-Pak® External Power Supply is an auxiliary part of EZ-Pak® Curve which can be used during routine filtration process.
Legal Information
EZ-PAK is a registered trademark of Merck KGaA, Darmstadt, Germany