Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CC127

Sigma-Aldrich

Caspase 2, recombinant human active

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
eCl@ss:
32160405

biological source

human

form

solid

specific activity

25 units

manufacturer/tradename

Chemicon®

technique(s)

activity assay: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... CASP2(835)

General description

Caspase-2 (also known as ICH-1, NEDD-2) is a member of the interleukin-1beta converting enzyme (ICE) family of cystein proteases. Similar as other caspases, caspase-2 also exists in cells as an inactive proenzyme. During apoptosis, procaspase-2 is processed at aspartate residues by self-proteolysis and/or cleavage by upstream caspases. The processed form of caspase-2 consists of large (19 kDa) and small (12 kDa) subunits which associate to form the active enzyme. The active recombinant human caspase-2 was expressed in E. coli. The expressed caspase-2 spontaneously undergoes autoprocessing to yield the subunits characteristic of the native enzyme. The recombinant caspase-2 preferentially cleaves the substrates consisting of consensus sequence VDVAD (e.g., VDVAD-AFC and VDVAD-pNA). The recombinant enzyme can also be used as a positive control in caspase assays or in determining the specificity of caspase substrates.Recommended usage: 0.5-1 unit for fluorometric caspase assays and 2-4 units for colorimetric caspase assays. Optimal working dilutions must be determined by end user.

Unit Definition

Specific Activity: Where one unit of the recombinant caspase-2 is the enzyme activity that cleaves 1 nmol of the caspase substrate VDVAD-pNA (pNA: pnitroanaline) per hour at 37oC in a reaction solution containing 50 mM Hepes, pH 7.2, 50 mM NaCl, 0.1% Chaps, 10 mM EDTA, 5% Glycerol, and 10 mM DTT.

Physical form

Partially purified recombinant caspase 2 with a full length HIS-6-tag. Approximately 10-20% caspase 2 protein. The remainder is E.Coli proteins. Approximately 0.8-2 mg total protein/U of activity; E.Coli proteins have no reactivity. Lyophilized. Reconstitute in 25 μL PBS containing 15% glycerol.

Storage and Stability

Maintain unopened and rehydrated product at -70°C in undiluted, freeze-thaw protected, tightly sealed aliquots. Avoid repeated freeze/thaw cycles. Product stable for 6 months from date of purchased if handled appropriately.

Analysis Note

Partially purified recombinant caspase 3 with a full length HIS-6-tag. Approximately 10-20% caspase 2 protein. The remainder is E.Coli proteins. Approximately 0.8-2 mg total protein/U of activity; E.Coli proteins have no reactivity.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The PIDDosome mediates delayed death of hippocampal CA1 neurons after transient global cerebral ischemia in rats.
Kuniyasu Niizuma,Hidenori Endo,Chikako Nito,D Jeannie Myer,Gab Seok Kim,Pak H Chan
Proceedings of the National Academy of Sciences of the USA null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service