Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BINDING-1

Millipore

VIRAPREP Binding Buffer

new

suitable for water monitoring, suitable for microbiological culture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161700
NACRES:
NA.74

form

liquid

shelf life

12 mo.

packaging

pack of 4 × 250 mL

technique(s)

microbiological culture: suitable

suitability

suitable for (Concentration of coliphage in water)

application(s)

water monitoring

storage temp.

2-8°C

General description

VIRAPREP Binding Buffer is a somatic coliphage and F-specific RNA bacteriophage concentration buffer designed to maintain bind and maintain viability of the microorganisms onto the filtration membrane.
It is a part of VIRAPREP Kit for the concentration of somatic coliphages and F-specific RNA bacteriophages.

Application

VIRAPREP Binding Buffer is used to concentrate somatic coliphages and F-specific RNA bacteriophages from drinking water samples.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service