Skip to Content
MilliporeSigma
All Photos(1)

Documents

ABX48

Sigma-Aldrich

Anti-Borrelia burgdorferi

Synonym(s):

Anti-Lyme Disease Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified antibody

clone

polyclonal

form

solution

species reactivity

all

packaging

pkg of 5 mL (ABX48-10KC)
pkg of 500 μL (ABX48-1KC)

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

relevant disease(s)

infectious diseases

shipped in

wet ice

storage temp.

2-8°C

Specificity

Reactive with WB bands present at 83kD, 41kD, 34kD,31kD and other low molecular weight bands. Cross reacts with Treponema pallidum, B. hermsii, and B. parkerii.

Immunogen

B. burgdoeferi whole cells

Application

This rabbit polyclonal Anti-Borrelia burgdorferi, Cat. No. ABX48, is tested for use in Immunohistochemistry, ELISA and Western Blotting.

Target description

Borrelia burgdorfer is a gram-negative spirochete, tick-borne obligate parasite whose normal reservoir is a variety of small mammals. It exists in North America and Europe and until 2016 was the only known cause of Lyme disease in North America (Borrelia mayonii, found in the midwestern US, is also known to cause the disease).

Physical form

Purified rabbit polyclonal in buffer containing 10 mM Phosphate Buffered Saline, pH 7.2 with 0.1% sodium azide.

Reconstitution

Please refer to Certificate of Analysis for concentration.

Storage and Stability

Store at 2-8 °C. For long-term storage, freeze at -20 °C

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service