Skip to Content
MilliporeSigma
All Photos(4)

Documents

ABS181

Sigma-Aldrich

Anti-PDE4B2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

cAMP-specific 3′,5′-cyclic phosphodiesterase 4B, DPDE4, PDE32

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

species reactivity (predicted by homology)

human (immunogen homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PDE4B(5142)

General description

Phosphodiesterase 4B2 (PDE4B2) is a member of the PDE cyclic nucleotides. PDE4B2 is thought to contain both UCR1 and UCR2 regulatory units. PDE4B2 has been associated with schizophrenia and depression and could be a promising target for autism therapies.

Specificity

This antibody recognizes PDE4B2 at the N-terminus.

Immunogen

Epitope: N-terminus
KLH-conjugated linear peptide corresponding to the N-terminus of human PDE4B2.

Application

Anti-PDE4B2 Antibody is an antibody against PDE4B2 for use in WB.
Research Category
Signaling
Research Sub Category
GPCR, cAMP/cGMP & Calcium Signaling

Quality

Evaluated by Western Blot in Rat brain membrane tissue lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected PDE4B2 on 10 µg of Rat brain membrane tissue lysate.

Target description

~78 kDa observed. UniProt describes 3 isoforms produced by alternative splicing: Isoform PDE4B1 at 83.3 kDa, Isoform PDE4B2 at 64.5 kDa, and Isoform PDE4B3 at 82.1 kDa

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Rat brain membrane tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anthony A Oliva et al.
Journal of neurochemistry, 123(6), 1019-1029 (2012-10-13)
Traumatic brain injury (TBI) results in significant inflammation which contributes to the evolving pathology. Previously, we have demonstrated that cyclic AMP (cAMP), a molecule involved in inflammation, is down-regulated after TBI. To determine the mechanism by which cAMP is down-regulated
Nicole M Wilson et al.
PloS one, 12(5), e0178013-e0178013 (2017-05-26)
Traumatic brain injury (TBI) initiates a deleterious inflammatory response that exacerbates pathology and worsens outcome. This inflammatory response is partially mediated by a reduction in cAMP and a concomitant upregulation of cAMP-hydrolyzing phosphodiesterases (PDEs) acutely after TBI. The PDE4B subfamily
Nicole M Wilson et al.
Frontiers in systems neuroscience, 10, 5-5 (2016-02-24)
Traumatic brain injury (TBI) results in significant impairments in hippocampal synaptic plasticity. A molecule critically involved in hippocampal synaptic plasticity, 3',5'-cyclic adenosine monophosphate, is downregulated in the hippocampus after TBI, but the mechanism that underlies this decrease is unknown. To

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service