Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABE1414

Sigma-Aldrich

Anti-MEIOB Antibody

serum, from rabbit

Synonym(s):

Meiosis-specific with OB domain-containing protein, Protein expressed in male leptotene and zygotene spermatocytes 675, MLZ-675

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$420.00

$420.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$420.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$420.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Meiob(75178)

General description

MEIOB is a single stranded binding protein required for proper homologous recombination in meiosis I. MEIOB is required for proper DNA strand repair at double strand breaks and faithful synapsis of the chromosomes. MEIOB is also required for successful completion of meiotic recombination and is part of a multiprotein complex with RPA2 and SPATA22 proteins. MEIOB is expressed in the gonads and specifically in ovary and testis starting at the 14th weeks post fertilization. MEIOB is also expressed in a variety of tumors but not in other tissues other than testis and ovary. Because of its unique expression MEIOB is being examined as a possible anti-cancer autoimmune agent.

Immunogen

Recombinant protein corresponding to mouse MEIOB.

Application

Immunoprecipitation Analysis: A representative lot immunoprecipitated MEIOB in P18 testes (Luo, M., et al. (2013). Nature. DOI: 10.1038/ncomms3788 pg. 1-12).
Immunocytochemistry Analysis: A representative lot detected MEIOB in mouse spermatocytes (Luo, M., et al. (2013). Nature. DOI: 10.1038/ncomms3788 pg. 1-12).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors
This Anti-MEIOB Antibody is validated for use in Western Blotting and Immunoprecipitation and Immunocytochemistry for the detection of MEIOB.

Quality

Evaluated by Western Blotting in mouse testis tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected MEIOB in 10 µg of mouse testis tissue lysate.

Target description

~50 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Rabbit polyclonal serum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kazumasa Takemoto et al.
Cell reports, 31(8), 107686-107686 (2020-05-28)
Meiotic recombination is critical for genetic exchange and generation of chiasmata that ensures faithful chromosome segregation during meiosis I. Meiotic recombination is initiated by DNA double-strand break (DSB) followed by multiple processes of DNA repair. The exact mechanisms for how
Jingjing Zhang et al.
Nature communications, 11(1), 2055-2055 (2020-04-30)
Breast cancer susceptibility gene II (BRCA2) is central in homologous recombination (HR). In meiosis, BRCA2 binds to MEILB2 to localize to DNA double-strand breaks (DSBs). Here, we identify BRCA2 and MEILB2-associating protein 1 (BRME1), which functions as a stabilizer of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service