Skip to Content
MilliporeSigma
All Photos(1)

Documents

AB5642P

Sigma-Aldrich

Anti-Cannabinoid Receptor 2 Antibody, CT

Chemicon®, from rabbit

Synonym(s):

CB2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Specificity

Recognizes rat Cannabinoid Receptor 2 (CB2). The immunogen shows no significant sequence homology with CB1.

SPECIES REACTIVITIES: The immunogen peptide is 88% conserved in mouse and 82% in human. Reactivity with other species has not been confirmed.

Immunogen

An 18 amino acid peptide sequence near the cytoplasmic, C-terminus of rat CB2 (Munro et al. 1993; Shire et al. 1996; Griffin et al. 2000).
Epitope: C-terminus

Application

Detect Cannabinoid Receptor 2 using this Anti-Cannabinoid Receptor 2 Antibody, C-terminus validated for use in ELISA & WB.
Research Category
Neuroscience
Research Sub Category
Neuroinflammation & Pain

Neurotransmitters & Receptors
Western blot: 1-10 μg/mL using ECL.

ELISA: 1:10,000-1:100,000 using 50-100 ng control peptide (Catalog Number AG214) per well.

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Liquid in phosphate buffered saline, pH 7.4, containing 0.1% BSA.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew P Ray et al.
Behavioural brain research, 196(1), 30-36 (2008-08-30)
Cisplatin chemotherapy frequently causes severe vomiting in two temporally separated clusters of bouts dubbed the acute and delayed phases. Cannabinoids can inhibit the acute phase, albeit through a poorly understood mechanism. We examined the substrates of cannabinoid-mediated inhibition of both

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service